Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gsdmcl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK019629.1 | - | 610 |
Gene Symbol: Gsdmcl-ps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK016931.1 | - | 657 |
Gene Symbol: Gsdmcl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK006248.1 | - | 4 | 617 |