Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Palm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028877.1 | NP_083153.1 | 7 | 1598 | ||
GenBank mRNA | BC113196.1 | - | 7 | 1601 | ||
BC113802.1 | - | 7 | 1596 | |||
EF208034.1 | - | 7 | 1598 |