Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cmip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163262.1 | NP_001156734.1 | 21 | 2656 | ||
NM_028941.1 | NP_083217.1 | 21 | 2479 |