Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ifrd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019242.1 | NP_062115.1 | 11, 12 | 1412 | ||
XM_008764643.1 | XP_008762865.1 | 10, 11 | 2031 | |||
GenBank mRNA | BC083560.1 | - | 11, 12 | 1478 | 7193329 | |
FQ222510.1 | - | 7, 8 | 836 | |||
J04511.1 | - | 11, 12 | 1412 |