Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wnk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290311.1 | NP_001277240.1 | 20 | 4489 | ||
NM_001290313.1 | NP_001277242.1 | 20 | 4489 | |||
NM_029361.4 | NP_083637.2 | 20 | 4489 | |||
GenBank mRNA | AK129439.1 | - | 9 | 1346 | ||
AK147550.1 | - | 20 | 4176 | |||
BC046464.1 | - | 13 | 2288 | |||
BC060187.1 | - | 9 | 1520 |