Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spaca3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029367.1 | NP_083643.1 | 2, 3 | 408 | ||
GenBank mRNA | AK006357.1 | - | 3, 4 | 556 | ||
AY572448.1 | - | 2, 3 | 371 | |||
AY601763.1 | - | 1, 2 | 91 | |||
BC100503.1 | - | 2, 3 | 408 | 30917230 |