Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cldn22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029383.1 | NP_083659.1 | 1 | 865 | ||
GenBank mRNA | AK008821.1 | - | 1 | 866 |