Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Egfem1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167748.1 | NP_001161220.1 | 13 | 2143 | ||
NM_029412.1 | NP_083688.1 | 15 | 2314 | |||
GenBank mRNA | AK032017.1 | - | 15 | 2158 | ||
CJ285936.1 | - | 1 | 204 |