siRNA Details
Gene Symbol: Ttc23l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029430.1 | NP_083706.1 | 11 | 1782 | ||
GenBank mRNA | AK015027.1 | - | 11 | 1754 |

Gene Symbol: Ttc23l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029430.1 | NP_083706.1 | 11 | 1782 | ||
GenBank mRNA | AK015027.1 | - | 11 | 1754 |