Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029655.3 | NP_083931.2 | 8 | 1219 | ||
GenBank mRNA | AK011015.1 | - | 8 | 1194 | ||
AK038971.1 | - | 8 | 1220 | |||
AK052487.1 | - | 8 | 1176 | |||
BC087942.1 | - | 8 | 1077 | 30303462 |