Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spcs3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029701.1 | NP_083977.1 | 5 | 1611 | ||
GenBank mRNA | AK037878.1 | - | 1 | 1006 | ||
AK048051.1 | - | 5 | 1598 | |||
BC019390.1 | - | 5 | 1540 | |||
BC054817.1 | - | 5 | 1576 | 5375997 |