Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arpc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029711.1 | NP_083987.1 | 10 | 1183 | ||
GenBank mRNA | AK083263.1 | - | 10 | 993 | ||
AK151513.1 | - | 10 | 1157 | |||
AK152356.1 | - | 11 | 1008 | |||
AK170061.1 | - | 11 | 982 | |||
BC099504.1 | - | 9 | 835 | |||
BC115750.1 | - | 10 | 981 | 40051600 |