Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trdn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029726.2 | NP_084002.2 | 38 | 3027 | ||
GenBank mRNA | AK036470.1 | - | 20 | 1567 | ||
AK085836.1 | - | 1 | 652 |