Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajc21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030046.2 | NP_084322.2 | 8 | 1150 | ||
GenBank mRNA | AK019636.1 | - | 3 | 245 | ||
BC048354.1 | - | 4 | 441 |