Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stxbp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081344.2 | NP_001074813.2 | 26 | 5357 | ||
GenBank mRNA | AK122417.1 | - | 5 | 2427 | ||
BC038042.1 | - | 6 | 2725 |