Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Naa11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033191.2 | NP_001028363.1 | 2 | 2153 | ||
GenBank mRNA | AK133248.1 | - | 2 | 2161 | ||
AK135863.1 | - | 2 | 2155 |