Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167883.1 | NP_001161355.1 | 4 | 2277 | ||
GenBank mRNA | AK129484.1 | - | 4 | 2262 | ||
AK220375.1 | - | 2 | 2414 |