Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FDFT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287742.1 | NP_001274671.1 | 3 | 797 | ||
NM_001287743.1 | NP_001274672.1 | 2 | 599 | |||
NM_001287745.1 | NP_001274674.1 | 1 | 308 | |||
NM_004462.4 | NP_004453.3 | 1 | 308 | |||
XM_017013217.1 | XP_016868706.1 | 2 | 598 | |||
GenBank mRNA | AK098682.1 | - | 882 | |||
AK296043.1 | - | 1 | 180 | |||
AK311246.1 | - | 1 | 173 | |||
AK315993.1 | - | 1 | 178 | |||
BC003573.1 | - | 1 | 155 | 3538137 | ||
BC009251.2 | - | 1 | 155 | 2961623 | ||
BC029641.1 | - | 1 | 118 | 4514761 | ||
BC034440.1 | - | 1 | 118 | |||
BG752446.1 | - | 172 | ||||
BI765996.1 | - | 3 | 401 | |||
BT006704.1 | - | 1 | 86 | |||
CR457033.1 | - | 1 | 86 | |||
DC314061.1 | - | 1 | 284 | |||
DC396859.1 | - | 1 | 310 | |||
L06070.1 | - | 1 | 95 | |||
L06105.1 | - | 1 | 130 | |||
S76822.1 | - | 1 | 151 | |||
X69141.1 | - | 1 | 177 |