Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc14a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019347.2 | NP_062220.2 | 5 | 717 | ||
NM_177962.3 | NP_808877.2 | 5 | 717 | |||
GenBank mRNA | AF031642.1 | - | 5 | 717 | ||
AF041788.1 | - | 5 | 717 | |||
AF042167.1 | - | 5 | 717 | |||
U77971.1 | - | 5 | 717 |