Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BUB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278616.1 | NP_001265545.1 | 24 | 3212 | ||
NM_001278617.1 | NP_001265546.1 | 24 | 3101 | |||
NM_004336.4 | NP_004327.1 | 25 | 3272 | |||
GenBank mRNA | AF011387.1 | - | 17 | 2337 | ||
AF043294.2 | - | 25 | 3192 | |||
AF046078.1 | - | 25 | 3209 | |||
AF047471.1 | - | 25 | 3195 | |||
AF053305.1 | - | 25 | 3223 | |||
AK225317.1 | - | 24 | 3057 | |||
AK291221.1 | - | 25 | 3225 | |||
AK302418.1 | - | 24 | 3168 | |||
BC028201.1 | - | 25 | 3193 | 5205385 |