Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pold1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021662.2 | NP_067694.1 | 1 | 126 | ||
XM_006229142.2 | XP_006229204.1 | 2 | 218 | |||
XM_006229143.2 | XP_006229205.1 | 2 | 1735 | |||
GenBank mRNA | AJ222691.1 | - | 1 | 130 |