Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adam3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020302.1 | NP_064698.1 | 2, 3 | 247 | ||
GenBank mRNA | BC078839.1 | - | 2, 3 | 153 | 7114315 | |
Y07903.1 | - | 2, 3 | 247 |