Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004075.4 | NP_004066.1 | 2 | 1084 | ||
XM_017018832.1 | XP_016874321.1 | 3 | 1219 | |||
GenBank mRNA | AK125915.1 | - | 4 | 747 | ||
AK290552.1 | - | 2 | 384 | |||
BC017466.1 | - | 2 | 507 | |||
BC030519.1 | - | 2 | 757 | 5169081 | ||
D83702.1 | - | 2 | 774 | |||
D84657.1 | - | 2 | 188 |