Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cep78 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021741.2 | NP_068509.2 | 5 | 752 | ||
XM_006231136.2 | XP_006231198.1 | 5 | 867 | |||
GenBank mRNA | FQ222521.1 | - | 3 | 617 | ||
X99330.1 | - | 5 | 540 |