Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ip6k2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021660.2 | NP_067692.2 | 4 | 503 | ||
XM_008766564.1 | XP_008764786.1 | 3 | 469 | |||
XM_008766565.1 | XP_008764787.1 | 2 | 179 | |||
XM_008766566.1 | XP_008764788.1 | 4 | 538 | |||
GenBank mRNA | BC083574.1 | - | 4 | 503 | 7112777 |