Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: S1pr5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021775.2 | NP_068543.2 | 3 | 838 | ||
XM_006242663.2 | XP_006242725.1 | 2 | 768 | |||
XM_006242664.2 | XP_006242726.1 | 2 | 1785 | |||
XM_006242665.2 | XP_006242727.1 | 2 | 1380 | |||
GenBank mRNA | AF115249.1 | - | 3 | 838 | ||
AF233649.1 | - | 1 | 581 | |||
FQ214303.1 | - | 2 | 685 |
Set Membership: |
G-protein coupled receptor |