Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ensa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033974.2 | NP_001029146.1 | 2 | 224 | ||
NM_021842.3 | NP_068614.1 | 2 | 224 | |||
GenBank mRNA | AJ005984.1 | - | 2 | 84 | ||
BC059135.1 | - | 2 | 228 | 6919652 | ||
CB812628.1 | - | 2 | 203 | |||
FQ212889.1 | - | 2 | 189 | |||
FQ212892.1 | - | 2 | 208 | |||
FQ212980.1 | - | 2 | 206 | |||
FQ213096.1 | - | 2 | 191 | |||
FQ213912.1 | - | 2 | 208 | |||
FQ214207.1 | - | 2 | 202 | |||
FQ216114.1 | - | 2 | 208 | |||
FQ216323.1 | - | 2 | 183 |