Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syt6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022191.1 | NP_071527.1 | 6 | 1792 | ||
GenBank mRNA | BC099185.1 | - | 1448 | 7383303 | ||
U20105.1 | - | 6 | 1792 |