Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gulo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022220.2 | NP_071556.2 | 12 | 1257 | ||
XM_006252146.2 | XP_006252208.1 | 11 | 1233 | |||
GenBank mRNA | BC089803.1 | - | 12 | 1257 | 7324344 | |
FQ219140.1 | - | 12 | 1282 | |||
FQ219632.1 | - | 12 | 1365 |