Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DUSP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004420.2 | NP_004411.2 | 7 | 2051 | ||
XM_011519932.2 | XP_011518234.1 | 7 | 3078 | |||
XM_011519933.2 | XP_011518235.1 | 7 | 2108 | |||
GenBank mRNA | BC038231.1 | - | 6 | 2054 | 5547764 | |
BC045110.1 | - | 6 | 1924 | 5478677 | ||
U27193.1 | - | 7 | 2057 |