Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RPLP0 SIRNA Assays ›
Gene Symbol:
Rplp0
Gene Name:
ribosomal protein, large, P0
Gene Aliases:
Arbp
Chromosome Location:
Chr.12: 46791528 - 46794797 on Build Rnor_6.0
UniGene ID:
Rn.1079
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
51868
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Rplp0
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_022402.2
NP_071797.1
4
529
GenBank mRNA
BC062028.1
-
3
498
5623881
DN931898.1
-
3
547
FQ209537.1
-
4
529
FQ209603.1
-
4
529
FQ209620.1
-
3
514
FQ209757.1
-
4
530
FQ210253.1
-
4
529
FQ210412.1
-
4
529
FQ211152.1
-
4
529
FQ211263.1
-
4
528
FQ212554.1
-
4
529
FQ212743.1
-
4
529
FQ212875.1
-
4
529
FQ212927.1
-
4
530
FQ213085.1
-
4
529
FQ213427.1
-
4
529
FQ213510.1
-
4
529
FQ213847.1
-
4
530
FQ214087.1
-
4
529
FQ214354.1
-
4
529
FQ214515.1
-
4
529
FQ214553.1
-
4
530
FQ214850.1
-
4
529
FQ214997.1
-
4
529
FQ215007.1
-
4
529
FQ215548.1
-
4
531
FQ215662.1
-
4
529
FQ215682.1
-
4
529
FQ215870.1
-
4
529
FQ216120.1
-
4
530
FQ216296.1
-
4
529
FQ216389.1
-
4
530
FQ216406.1
-
4
532
FQ216495.1
-
4
529
FQ216618.1
-
4
529
FQ216655.1
-
4
530
FQ216733.1
-
4
531
FQ216774.1
-
4
529
FQ216813.1
-
4
528
FQ216851.1
-
4
529
FQ216913.1
-
4
530
FQ217406.1
-
4
529
FQ218561.1
-
4
529
FQ218832.1
-
4
529
FQ218943.1
-
4
529
FQ219025.1
-
4
532
FQ219184.1
-
4
533
FQ219389.1
-
4
529
FQ219457.1
-
4
529
FQ219678.1
-
4
529
FQ219815.1
-
4
529
FQ219817.1
-
4
529
FQ219837.1
-
4
529
FQ219855.1
-
4
529
FQ219923.1
-
4
529
FQ220008.1
-
4
529
FQ220051.1
-
4
529
FQ220077.1
-
4
529
FQ220115.1
-
4
529
FQ220181.1
-
4
529
FQ220201.1
-
4
527
FQ220213.1
-
4
529
FQ220247.1
-
4
530
FQ220282.1
-
4
531
FQ220284.1
-
4
529
FQ220302.1
-
4
529
FQ220338.1
-
4
529
FQ220345.1
-
4
529
FQ220370.1
-
4
529
FQ220406.1
-
4
529
FQ220413.1
-
4
529
FQ220420.1
-
4
530
FQ220433.1
-
4
526
FQ220444.1
-
4
529
FQ220445.1
-
4
525
FQ220449.1
-
4
529
FQ220500.1
-
4
529
FQ220514.1
-
4
529
FQ220527.1
-
4
529
FQ220533.1
-
4
561
FQ220542.1
-
4
529
FQ220545.1
-
4
529
FQ220634.1
-
4
529
FQ220654.1
-
4
529
FQ220684.1
-
4
530
FQ220690.1
-
4
531
FQ220769.1
-
4
529
FQ220787.1
-
4
529
FQ220801.1
-
4
529
FQ220833.1
-
4
529
FQ220838.1
-
4
529
FQ220888.1
-
4
529
FQ220932.1
-
4
530
FQ221146.1
-
4
529
FQ221380.1
-
4
529
FQ221533.1
-
4
530
FQ221556.1
-
4
527
FQ222171.1
-
4
529
FQ222181.1
-
4
529
FQ222327.1
-
4
529
FQ222772.1
-
4
529
FQ222906.1
-
4
529
FQ223121.1
-
4
529
FQ223487.1
-
4
529
FQ224547.1
-
4
529
FQ224834.1
-
4
529
FQ224846.1
-
4
529
FQ225170.1
-
4
529
FQ225608.1
-
4
529
FQ225654.1
-
4
531
FQ225860.1
-
4
531
FQ225886.1
-
4
529
FQ226306.1
-
4
531
FQ227381.1
-
4
529
FQ227522.1
-
4
524
FQ227719.1
-
4
529
FQ227837.1
-
4
529
FQ227937.1
-
4
530
FQ227941.1
-
4
529
FQ227942.1
-
4
649
FQ228548.1
-
4
529
FQ228995.1
-
4
529
FQ228996.1
-
4
529
FQ229139.1
-
4
531
FQ229159.1
-
4
529
FQ229164.1
-
4
530
FQ229208.1
-
4
529
FQ229241.1
-
4
529
FQ229244.1
-
4
529
FQ229248.1
-
2
409
FQ229272.1
-
4
529
FQ229273.1
-
4
530
FQ229303.1
-
4
529
FQ229337.1
-
4
529
FQ229352.1
-
4
529
FQ229365.1
-
4
529
FQ229397.1
-
4
529
FQ229408.1
-
4
530
FQ229433.1
-
4
529
FQ229448.1
-
4
529
FQ229521.1
-
4
529
FQ229547.1
-
4
529
FQ229554.1
-
4
529
FQ229566.1
-
4
525
FQ229610.1
-
4
529
FQ229626.1
-
4
526
FQ229726.1
-
4
529
FQ229740.1
-
4
529
FQ229742.1
-
4
529
FQ229776.1
-
4
529
FQ229808.1
-
4
529
FQ229815.1
-
4
529
FQ229822.1
-
4
529
FQ229830.1
-
4
531
FQ229883.1
-
4
531
FQ229901.1
-
4
529
FQ229929.1
-
4
529
FQ229932.1
-
2
427
FQ229952.1
-
4
529
FQ229974.1
-
4
529
FQ230005.1
-
4
529
FQ230055.1
-
4
528
FQ230127.1
-
4
529
FQ230267.1
-
4
532
FQ230943.1
-
4
529
FQ231445.1
-
4
531
FQ231525.1
-
4
529
FQ231591.1
-
4
531
FQ231710.1
-
4
530
FQ231733.1
-
4
530
FQ231897.1
-
4
531
FQ231967.1
-
4
529
FQ232029.1
-
4
530
FQ232154.1
-
4
529
FQ232165.1
-
4
529
FQ232310.1
-
4
528
FQ232319.1
-
4
530
FQ232697.1
-
4
531
FQ232767.1
-
4
529
FQ232951.1
-
4
529
FQ233152.1
-
4
528
FQ233377.1
-
4
529
FQ233473.1
-
4
529
FQ233732.1
-
4
529
FQ233808.1
-
4
529
FQ234195.1
-
4
529
FQ234341.1
-
4
529
FQ234613.1
-
4
529
FQ234733.1
-
4
529
FQ234735.1
-
4
419
FQ234863.1
-
4
529
FQ235063.1
-
4
529
FQ235079.1
-
4
529
FQ235122.1
-
4
529
FQ235259.1
-
4
529
X15096.1
-
4
529
Z29530.1
-
3
485
Back To Top
More Information
Panther Classification:
Molecular Function -
ribosomal protein
translational protein
Gene Ontology Categories:
Function(s)
Process(es)
translational elongation
ribosome biogenesis
cellular response to interleukin-4
structural constituent of ribosome
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-jmrqp:80/10.239.165.107:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0