Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tep1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022591.2 | NP_072113.1 | 2 | 680 | ||
GenBank mRNA | U89282.1 | - | 2 | 681 |