Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prpsap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022545.3 | NP_071990.2 | 5 | 671 | ||
XM_008768391.1 | XP_008766613.1 | 5 | 608 | |||
GenBank mRNA | BC078822.1 | - | 5 | 646 | 7128451 | |
D26073.1 | - | 5 | 569 |