Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022798.2 | NP_073635.1 | 2 | 209 | ||
GenBank mRNA | AF156272.1 | - | 2 | 316 | ||
BC078819.1 | - | 2 | 209 | 7113511 |