Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DEDD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039711.1 | NP_001034800.1 | 5 | 1889 | ||
NM_001039712.1 | NP_001034801.1 | 6 | 1860 | |||
NM_032998.2 | NP_127491.1 | 6 | 1922 | |||
XM_005245597.3 | XP_005245654.1 | 6 | 1923 | |||
XM_005245598.4 | XP_005245655.1 | 6 | 3062 | |||
XM_005245599.2 | XP_005245656.1 | 6 | 1966 | |||
XM_005245600.2 | XP_005245657.1 | 5 | 2074 | |||
XM_017002802.1 | XP_016858291.1 | 6 | 2972 | |||
GenBank mRNA | AK001497.1 | - | 6 | 1825 | ||
AK022531.1 | - | 7 | 1745 | |||
BC013910.2 | - | 6 | 1902 | 3504847 | ||
BC016724.1 | - | 6 | 1823 | 4077893 |