Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptprq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022925.1 | NP_075214.1 | 3 | 535 | ||
GenBank mRNA | AF063249.1 | - | 3 | 535 |
Gene Symbol: LOC102552564 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006241319.2 | XP_006241381.1 | 3 | 539 |
Set Membership: |
G-protein coupled receptor |