Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPHA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304536.1 | NP_001291465.1 | 3, 4 | 440 | ||
NM_004438.4 | NP_004429.1 | 2, 3 | 275 | |||
XM_005246374.2 | XP_005246431.1 | 2, 3 | 275 | |||
GenBank mRNA | AK225181.1 | - | 2, 3 | 221 | ||
AK290306.1 | - | 3, 4 | 440 | |||
AK312380.1 | - | 2, 3 | 221 | |||
BC026327.1 | - | 2, 3 | 200 | 4814429 | ||
BC105000.1 | - | 2, 3 | 201 | 8144003 | ||
BC105002.1 | - | 2, 3 | 201 | 8144005 | ||
DA451606.1 | - | 2, 3 | 275 | |||
L36645.1 | - | 2, 3 | 198 |