Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mcam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034009.1 | NP_001029181.1 | 3 | 233 | ||
NM_023983.3 | NP_076473.2 | 3 | 233 | |||
GenBank mRNA | AB035506.1 | - | 3 | 231 | ||
AB035507.1 | - | 3 | 231 | |||
BC070916.1 | - | 3 | 233 | 7105099 |