Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpina5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244739.1 | NP_001231668.1 | 2 | 425 | ||
NM_022957.3 | NP_075246.3 | 2 | 483 | |||
XM_006240494.2 | XP_006240556.1 | 5 | 2598 | |||
GenBank mRNA | AB013128.1 | - | 2 | 356 | ||
BC081797.1 | - | 2 | 393 | 7112069 | ||
BC097315.1 | - | 2 | 440 | 7453034 | ||
CK603584.1 | - | 2 | 425 |