Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asic1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024154.2 | NP_077068.1 | 5 | 860 | ||
XM_006257378.2 | XP_006257440.1 | 3 | 1185 | |||
XM_006257381.2 | XP_006257443.1 | 5 | 1145 | |||
XM_008765793.1 | XP_008764015.1 | 3 | 1189 | |||
XM_008765794.1 | XP_008764016.1 | 5 | 1145 | |||
XM_008765795.1 | XP_008764017.1 | 5 | 798 | |||
XM_008765796.1 | XP_008764018.1 | 5 | 1145 | |||
GenBank mRNA | AB049451.1 | - | 3 | 451 | ||
AJ006519.1 | - | 3 | 906 | |||
AJ309926.1 | - | 3 | 910 |