Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Axin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024405.1 | NP_077381.1 | 308 | |||
GenBank mRNA | AF017756.1 | - | 308 |