Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc21a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030837.1 | NP_110464.1 | 8 | 770 | ||
GenBank mRNA | AB012662.1 | - | 7 | 659 | ||
AF445993.1 | - | 6 | 612 | |||
AF445994.1 | - | 6 | 565 | |||
AF445995.1 | - | 7 | 698 | |||
AF445996.1 | - | 6 | 565 | |||
AF445997.1 | - | 7 | 698 | |||
AF445998.1 | - | 6 | 612 | |||
AF445999.1 | - | 5 | 479 | |||
AF446000.1 | - | 4 | 337 | |||
AF446001.1 | - | 4 | 363 | |||
AF446002.1 | - | 5 | 470 | |||
AF446003.1 | - | 4 | 337 | |||
AF446004.1 | - | 4 | 321 | |||
AF446005.1 | - | 4 | 337 | |||
AF446006.1 | - | 5 | 470 | |||
D79981.1 | - | 8 | 770 |