Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnai2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031035.3 | NP_112297.1 | 1 | 259 | ||
XM_006243858.1 | XP_006243920.1 | 1 | 289 | |||
GenBank mRNA | AY899210.1 | - | 1 | 107 | ||
AY899211.1 | - | 1 | 110 | |||
BC078806.1 | - | 1 | 211 | 7121183 | ||
CO394955.1 | - | 1 | 259 | |||
DQ120471.1 | - | 1 | 168 | |||
DQ120472.1 | - | 1 | 168 | |||
M12672.1 | - | 1 | 155 | |||
M17528.1 | - | 1 | 257 |