Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030864.1 | NP_110491.1 | 2 | 434 | ||
GenBank mRNA | AF034214.1 | - | 2 | 434 |