Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPARA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001928.2 | NP_001001928.1 | 3 | 319 | ||
NM_005036.4 | NP_005027.2 | 4 | 403 | |||
XM_005261655.3 | XP_005261712.1 | 4 | 696 | |||
XM_005261656.3 | XP_005261713.1 | 5 | 860 | |||
XM_006724269.3 | XP_006724332.1 | 4 | 462 | |||
XM_006724270.3 | XP_006724333.1 | 3 | 368 | |||
XM_011530239.2 | XP_011528541.1 | 5 | 552 | |||
XM_011530240.2 | XP_011528542.1 | 5 | 555 | |||
XM_011530241.2 | XP_011528543.1 | 4 | 460 | |||
XM_011530242.2 | XP_011528544.1 | 3 | 282 | |||
XM_011530243.2 | XP_011528545.1 | 2 | 523 | |||
XM_011530244.2 | XP_011528546.1 | 5 | 555 | |||
XM_011530245.2 | XP_011528547.1 | 3 | 371 | |||
XM_017028839.1 | XP_016884328.1 | 5 | 552 | |||
XM_017028840.1 | XP_016884329.1 | 5 | 555 | |||
GenBank mRNA | AB307690.1 | - | 1 | 137 | ||
AK289821.1 | - | 4 | 696 | |||
BC000052.2 | - | 3 | 347 | |||
CR456547.1 | - | 2 | 273 | |||
CR457435.1 | - | 1 | 137 | |||
EU395809.1 | - | 1 | 137 | |||
EU650667.1 | - | 1 | 137 | |||
HQ692862.1 | - | 1 | 137 | |||
L02932.1 | - | 3 | 353 | |||
S74349.1 | - | 2 | 260 | |||
Y07619.1 | - | 1 | 137 |