Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nell2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031070.2 | NP_112332.2 | 6, 7 | 692 | ||
XM_008765797.1 | XP_008764019.1 | 5, 6 | 1042 | |||
XM_008765798.1 | XP_008764020.1 | 6, 7 | 663 | |||
GenBank mRNA | AY089719.1 | - | 6, 7 | 692 | ||
BC093617.1 | - | 5, 6 | 636 | 7312010 | ||
EF110908.1 | - | 4, 5 | 485 | |||
GQ376510.1 | - | 5, 6 | 614 | |||
U48245.1 | - | 6, 7 | 692 |