Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005050.3 | NP_005041.1 | 2 | 247 | ||
NR_003256.2 | 2 | 247 | ||||
XM_005267939.3 | XP_005267996.1 | 2 | 180 | |||
XM_005267941.4 | XP_005267998.1 | 2 | 142 | |||
XM_005267949.3 | XP_005268006.1 | 2 | 245 | |||
XM_011537041.2 | XP_011535343.1 | 2 | 190 | |||
XM_017021531.1 | XP_016877020.1 | 2 | 180 | |||
XM_017021532.1 | XP_016877021.1 | 2 | 180 | |||
XM_017021533.1 | XP_016877022.1 | 2 | 180 | |||
XM_017021535.1 | XP_016877024.1 | 2 | 180 | |||
XM_017021536.1 | XP_016877025.1 | 2 | 128 | |||
XM_017021537.1 | XP_016877026.1 | 2 | 151 | |||
XM_017021539.1 | XP_016877028.1 | 2 | 117 | |||
XM_017021541.1 | XP_016877030.1 | 2 | 117 | |||
XM_017021544.1 | XP_016877033.1 | 2 | 180 | |||
GenBank mRNA | AF009746.1 | - | 2 | 146 | ||
AK291332.1 | - | 2 | 117 | |||
AK308382.1 | - | 2 | 128 | |||
BC012815.2 | - | 2 | 101 | 2960427 | ||
BI916327.1 | - | 2 | 239 | |||
BT007412.1 | - | 2 | 95 | |||
BX248053.1 | - | 2 | 175 | |||
CR457104.1 | - | 2 | 95 | |||
DA713794.1 | - | 2 | 181 | |||
DC332064.1 | - | 2 | 247 |