Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMAD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005902.3 | NP_005893.1 | 1 | 387 | ||
XM_011521559.2 | XP_011519861.1 | 1 | 546 | |||
GenBank mRNA | AI762628.1 | - | 1 | 387 | ||
AK290881.1 | - | 2 | 312 | |||
BC050743.1 | - | 1 | 305 | 6094328 | ||
BF058230.1 | - | 1 | 387 | |||
U68019.1 | - | 1 | 155 | |||
U76622.1 | - | 1 | 126 |