Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cldn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031699.2 | NP_113887.2 | 2 | 504 | ||
GenBank mRNA | AF195500.1 | - | 2 | 445 | ||
BC061992.1 | - | 2 | 504 | 5599462 | ||
FQ220868.1 | - | 2 | 473 |