Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031706.1 | NP_113894.1 | 3 | 179 | ||
GenBank mRNA | BC082802.1 | - | 2 | 163 | 7302521 | |
BC166878.1 | - | 3 | 179 | 6920981 | ||
FQ221315.1 | - | 3 | 179 | |||
FQ222655.1 | - | 3 | 216 | |||
FQ222951.1 | - | 3 | 177 | |||
FQ223011.1 | - | 3 | 179 | |||
FQ223286.1 | - | 3 | 181 | |||
FQ229267.1 | - | 3 | 179 | |||
X06423.1 | - | 3 | 179 |
Gene Symbol: LOC297756 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013928.2 | NP_001013950.2 | 1 | 153 | ||
GenBank mRNA | AY383664.1 | - | 1 | 153 |